Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyldiphenylphosphine, 98%

Catalog No. p-7024095
Click to view available options
Quantity:
10 g
50 g

CAS: 607-01-2 | C14H15P | 214.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-01-2
Molecular Formula C14H15P
Molecular Weight (g/mol) 214.25
MDL Number MFCD00015170
InChI Key WUOIAOOSKMHJOV-UHFFFAOYSA-N
Synonym ethyldiphenylphosphine, phosphine, ethyldiphenyl, diphenyl ethyl phosphine, diphenylphosphinoethane, diphenylphosphinoethan, ethyl diphenylphosphine, ethyl diphenyl phosphane, acmc-209mlk, ethyl diphenyl phosphine
PubChem CID 69082
IUPAC Name ethyl(diphenyl)phosphane
SMILES CCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.048
Boiling Point 130°C to 132°C (1 mmHg)
Odor Stench
Quantity 10 g
UN Number UN3278
Sensitivity Air Sensitive
Formula Weight 214.25
Percent Purity 98%
Chemical Name or Material Ethyldiphenylphosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.