Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2,4-dioxovalerate, 98%, Thermo Scientific Chemicals

Catalog No. p-3757853
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-79-2
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.15
MDL Number MFCD00009124
InChI Key OYQVQWIASIXXRT-UHFFFAOYSA-N
Synonym ethyl 2,4-dioxovalerate, ethyl acetopyruvate, pentanoic acid, 2,4-dioxo-, ethyl ester, ethyl2,4-dioxovalerate, 2,4-dioxo-pentanoic acid ethyl ester, 2,4-dioxopentanoic acid ethyl ester, 2,4-dioxo-pentanoic-acid-ethyl-ester, 2,4-dioxo-pentanoicacid ethylester, ethyl 2,4-dioxo-valerate, ethyl 2,4-dioxo-pentanoate
PubChem CID 69208
IUPAC Name ethyl 2,4-dioxopentanoate
SMILES CCOC(=O)C(=O)CC(C)=O

Specifications

Melting Point 16.0°C to 18.0°C
Color Yellow
Density 1.1200g/mL
Boiling Point 101.0°C to 103.0°C (12.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4730 to 1.4750
Linear Formula CH3COCH2COCO2C2H5
Quantity 25 g
Beilstein 03, 747
Specific Gravity 1.12
Formula Weight 158.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 2,4-dioxovalerate
Show More Show Less
EINECSNumber 210-447-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.