Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 2-cyanoacrylate

Catalog No. ac434450050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 7085-85-0 | C6H7NO2 | 125.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7085-85-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
MDL Number MFCD00045615
InChI Key FGBJXOREULPLGL-UHFFFAOYSA-N
Synonym ethyl 2-cyanoacrylate, ethyl cyanoacrylate, 2-propenoic acid, 2-cyano-, ethyl ester, unii-2g95foh7sf, ccris 1693, acrylic acid, 2-cyano-, ethyl ester, 2g95foh7sf, 2-cyanoacrylic acid ethyl ester, 2-propenoic acid,2-cyano-, ethyl ester, dsstox_cid_5279
PubChem CID 81530
IUPAC Name ethyl 2-cyanoprop-2-enoate
SMILES CCOC(=O)C(=C)C#N

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 54.0°C to 56.0°C
Flash Point 85°C
Packaging Glass bottle
Linear Formula CNC(=CH2)COOCH2CH3
Quantity 5 g
Specific Gravity 1.04
Merck Index 15,3842
Solubility Information Solubility in water: reacts. Other solubilities: soluble in methyl ethyl ketone, toluene, acetone,, dmf, nitromethane
Formula Weight 125.13
Concentration or Composition (by Analyte or Components) may contain varying amounts of polymerized material
Physical Form Liquid
Chemical Name or Material Ethyl 2-cyanoacrylate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 230-391-5
Recommended Storage −20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.