Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle3-aminobenzoate, +99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
582-33-2
C9H11NO2
165.19
MFCD00007794
ZMCBYSBVJIMENC-UHFFFAOYSA-N
3-aminobenzoic acid ethyl ester,m-ethoxycarbonylaniline,3-ethoxycarbonyl aniline,ethyl m-aminobenzoate,benzoic acid, 3-amino-, ethyl ester,m-aminobenzoic acid, ethyl ester,m-aminobenzoate,metacaine,3-amino-benzoic acid ethyl ester,benzoic acid, m-amino-, ethyl ester
11400
CHEBI:81494
ethyl 3-aminobenzoate
CCOC(=O)C1=CC(=CC=C1)N
582-33-2
C9H11NO2
165.19
MFCD00007794
ZMCBYSBVJIMENC-UHFFFAOYSA-N
3-aminobenzoic acid ethyl ester,m-ethoxycarbonylaniline,3-ethoxycarbonyl aniline,ethyl m-aminobenzoate,benzoic acid, 3-amino-, ethyl ester,m-aminobenzoic acid, ethyl ester,m-aminobenzoate,metacaine,3-amino-benzoic acid ethyl ester,benzoic acid, m-amino-, ethyl ester
11400
CHEBI:81494
ethyl 3-aminobenzoate
CCOC(=O)C1=CC(=CC=C1)N
Specific Gravity 1.107
Boiling Point 172°C to 175°C (13.0mmHg)
Flash Point >112°C
Refractive Index 1.5595 to 1.5615
Packaging Glass bottle
Color Brown to Yellow
Quantity 5g
Infrared Spectrum Authentic
Density 1.1070g/mL
Assay Percent Range 99+%
Linear Formula H2NC6H4CO2C2H5
Beilstein 14, 389
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol,diethyl ether,soluble in chloroform
Formula Weight 165.19
Physical Form Liquid or Low Melting Solid
Percent Purity ≥99.0%
Chemical Name or Material Ethyl 3-aminobenzoate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning