Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle3-ethoxybut-2-enoate, 97%, Thermo Scientific™

Catalog No. CD10992EA
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 998-91-4
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00026932,MFCD00026932,MFCD00026932
InChI Key ZOCYCSPSSNMXBU-VOTSOKGWSA-N
Synonym ethyl 3-ethoxycrotonate, crotonic acid, 3-ethoxy-, ethyl ester, ethyl 2z-3-ethoxybut-2-enoate, ethyl 3-ethoxybut-2-enoate, 3-ethoxy-2-butenoic acid ethyl ester, ethyl beta-ethoxycrotonate, 3-ethoxycrotonic acid ethyl ester, ethyl z-3-ethoxybut-2-enoate, z-3-ethoxy-but-2-enoic acid ethyl ester
PubChem CID 5904488
IUPAC Name ethyl (2E)-3-ethoxybut-2-enoate
SMILES CCO\C(C)=C\C(=O)OCC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2OC(CH3)=CHCO2CH2CH3
Quantity 10 g
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-ethoxybut-2-enoate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.