Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 4-cyanobenzoate, 99%, Thermo Scientific™

Catalog No. p-2691444
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7153-22-2
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00001824
InChI Key JLSSWDFCYXSLQX-UHFFFAOYSA-N
Synonym benzoic acid, 4-cyano-, ethyl ester, 4-cyanobenzoic acid ethyl ester, benzoic acid, 4-cyano-,ethyl ester, 4-cyano-benzoic acid ethyl ester, ethyl p-cyanobenzoate, ethyl-4-cyanobenzoate, p-cyano ethyl benzoate, acmc-209ypo, ethyl 4-cyanobenzoate, 4-cyanobenzoic acid, ethyl ester
PubChem CID 81589
IUPAC Name ethyl 4-cyanobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 52°C to 55°C
Flash Point >110°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula NCC6H4CO2C2H5
Quantity 5 g
Beilstein 09, 846
Formula Weight 175.19
Percent Purity 99%
Chemical Name or Material Ethyl 4-cyanobenzoate, 99%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-500-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.