Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylaminoacetaldehyde dimethylacetal, 97%

Catalog No. AC390910050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 38711-20-5 | C6H15NO2 | 133.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38711-20-5
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.19
MDL Number MFCD00010205
InChI Key HUYAEQCJNXODLQ-UHFFFAOYSA-N
Synonym ethanamine, 2,2-dimethoxy-n,n-dimethyl, dimethylamino acetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethyl acetal, dimethylaminoacetaldehyde dimethylacetal, n-2,2-dimethoxyethyl dimethylamine, 2-dimethylamino acetaldehyde dimethylacetal, 2,2-dimethoxyethyl dimethylamine, 2,2-dimethoxyethyl dimethyl amine, 2,2-dimethoxy-n,n-dimethylethanamin, acmc-209j1l
PubChem CID 123476
IUPAC Name 2,2-dimethoxy-N,N-dimethylethanamine
SMILES COC(CN(C)C)OC

Specifications

Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 64°C to 66°C (12.0 mmHg)
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.4100 to 1.412
Linear Formula (CH3)2NCH2CH(OCH3)2
Quantity 5 g
Specific Gravity 0.9
Formula Weight 133.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylaminoacetaldehyde dimethylacetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 254-097-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.