Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl caproate, 99+%

Catalog No. p-3757828
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle

CAS: 123-66-0 | C8H16O2 | 144.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-66-0
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009511
InChI Key SHZIWNPUGXLXDT-UHFFFAOYSA-N
Synonym ethyl caproate, ethyl hexoate, hexanoic acid, ethyl ester, hexanoic acid ethyl ester, capronic ether absolute, caproic acid ethyl ester, ethyl n-hexanoate, ethyl butyl acetate, caproate, acetic acid, butyl-, ethyl ester
PubChem CID 31265
ChEBI CHEBI:86055
IUPAC Name ethyl hexanoate
SMILES CCCCCC(=O)OCC

Specifications

Acidity 1mg KOH/g max.
Melting Point 67°C
Color Colorless
Density 0.8690g/mL
Boiling Point 168°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.409
Linear Formula CH3(CH2)4CO2C2H5
Quantity 5 mL
Beilstein 02, 323
Specific Gravity 0.869
Merck Index 15, 3832
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in glycerol,soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl caproate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-640-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.