Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium hydroxide, 25% in water

Catalog No. AC420520010
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle

CAS: 75-59-2 | C4H13NO | 91.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 25% in Water
CAS 75-59-2, 7732-18-5
Melting Point -25.0°C
Color Colorless to Yellow
Density 1.0140g/mL
Boiling Point 102.0°C
Flash Point >95°C
Packaging Plastic bottle
Molecular Formula C4H13NO
Refractive Index 1.3780 to 1.3840 (20°C, 589nm)
Linear Formula (CH3)4NOH
Quantity 1 kg
MDL Number MFCD00008280
Beilstein 04,5
Fieser 11,514
Specific Gravity 1.014
Merck Index 15,9371
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
Solubility Information Solubility in water: soluble.
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES C[N+](C)(C)C.[OH-]
IUPAC Name tetramethylazanium;hydroxide
Molecular Weight (g/mol) 91.15
PubChem CID 60966
Viscosity 3.13 mPa.s (19°C)
Formula Weight 91.15
Percent Purity 23 to 27%
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.