Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl isothiocyanate, 96%

Catalog No. AC376860250
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle

CAS: 542-85-8 | C3H5NS | 87.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-85-8
Molecular Formula C3H5NS
Molecular Weight (g/mol) 87.14
MDL Number MFCD00004820
InChI Key HBNYJWAFDZLWRS-UHFFFAOYSA-N
Synonym ethyl isothiocyanate, ethane, isothiocyanato, ethyl mustard oil, ethylisothiocyanate, isothiocyanic acid, ethyl ester, unii-3284mj2t8p, ccris 7323, ethylisothio-cyanate, isothiocyanato-ethane
PubChem CID 10966
ChEBI CHEBI:85098
IUPAC Name isothiocyanatoethane
SMILES CCN=C=S

Specifications

Melting Point -6°C
Color Colorless to Yellow
Density 0.9950g/mL
Boiling Point 130°C to 132°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5100 to 1.513
Linear Formula C2H5NCS
Quantity 25 g
Specific Gravity 0.995
Merck Index 15, 3870
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
Formula Weight 87.14
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Ethyl isothiocyanate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Causes serious eye damage.
Toxic in contact with skin.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-831-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.