Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl N-(diphenylmethylene)glycinate, 98%

Catalog No. p-3757847
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

CAS: 69555-14-2 | C17H17NO2 | 267.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69555-14-2
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.32
MDL Number MFCD00010590
InChI Key QUGJYNGNUBHTNS-UHFFFAOYSA-N
Synonym ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
PubChem CID 319508
IUPAC Name ethyl 2-(benzhydrylideneamino)acetate
SMILES CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 51°C to 55°C
Color White to Yellow
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2C=NCH2COOCH2CH3
Quantity 25 g
Formula Weight 267.32
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Ethyl N-(diphenylmethylene)glycinate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.