Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene carbonate, 99+%

Catalog No. p-3736441
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum

CAS: 96-49-1 | C3H4O3 | 88.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-49-1
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.06
MDL Number MFCD00005382
InChI Key KMTRUDSVKNLOMY-UHFFFAOYSA-N
Synonym ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec
PubChem CID 7303
IUPAC Name 1,3-dioxolan-2-one
SMILES C1COC(=O)O1

Specifications

Melting Point 34.0°C to 37.0°C
Color White to Yellow
Density 1.3210g/mL
Boiling Point 246.7°C
Flash Point 150°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Quantity 250 g
Beilstein 19, 100
Fieser 01,372
Specific Gravity 1.321
Solubility Information Solubility in water: 214g/L (20°C). Other solubilities: soluble in ether,n-butanol and carbon tetra-,chloride,miscible with alcohol,ethyl acetate,benzene and,chloroform
Viscosity 1.92 mPa.s (40°C)
Formula Weight 88.06
Percent Purity 99+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Ethylene carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 202-510-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.