Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium hydroxide pentahydrate, 98%

Catalog No. AC420530250
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Melting Point 62°C to 71°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula (CH3)4NOH·5H2O
Quantity 25 g
Beilstein 04, 50
Solubility Information Solubility in water: soluble
Formula Weight 181.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life w
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.