Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Bis(3-aminopropyl) Ether 98.0+%, TCI America™
SDP

Catalog No. D184025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184025ML 25 mL
D18405ML 5 mL
D1840500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D184025ML Supplier TCI America Supplier No. D184025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2997-01-5
Molecular Formula C8H22N2O2
Molecular Weight (g/mol) 178.28
MDL Number MFCD00059938
InChI Key POTQBGGWSWSMCX-UHFFFAOYSA-P
Synonym di 3-aminopropoxy ethane, 1-propanamine, 3,3'-1,2-ethanediylbis oxy bis, 1,2-bis 3-aminopropoxy ethane, 3,3'-ethane-1,2-diylbis oxy bis propan-1-amine, 3,3'-ethylenedioxy bispropylamine, 3,3'-ethylenedioxybis propylamine, propylamine, 3,3'-ethylenedioxy bis, ethylene glycol bis 3-aminopropyl ether, 1,10-diamino-4,7-dioxadecane, 3-2-3-aminopropoxy ethoxy propan-1-amine
PubChem CID 76342
IUPAC Name 3-[2-(3-aminopropoxy)ethoxy]propan-1-amine
SMILES C(CN)COCCOCCCN

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 mL
UN Number 3267
Formula Weight 176.26
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Bis(3-aminopropyl) Ether
RTECSNumber UI2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.