Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylene Glycol (Certified), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Glass Bottle
HDPE Bottle
Lined Steel Drum
Specifications
Specifications
pH | 5.5 to 7.5 |
Melting Point | -13°C |
Color | Colorless |
Acidity | 0.01% max. |
Decomposition Information | >500°C |
Density | 1.113g/cm³ |
Boiling Point | 196°C |
Quantity | 500 mL |
Flash Point | 116°C (240°F) |
Identification | Pass Test |
Show More |
Safety and Handling

Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:1 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction