Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol diacetate, 99%

Catalog No. p-3778568
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum

CAS: 111-55-7 | C6H10O4 | 146.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-55-7
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00008718
InChI Key JTXMVXSTHSMVQF-UHFFFAOYSA-N
Synonym ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus
PubChem CID 8121
IUPAC Name 2-acetyloxyethyl acetate
SMILES CC(=O)OCCOC(=O)C

Specifications

Melting Point -41°C
Density 1.1200g/mL
Boiling Point 186°C to 187°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4142 to 1.4162
Linear Formula CH3CO2CH2CH2O2CCH3
Quantity 500 mL
Beilstein 02, 142
Specific Gravity 1.12
Merck Index 15, 3853
Solubility Information Solubility in water: 160g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene,miscible with ethanol and acetone
Viscosity 2.58 mm2/s (20°C)
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethylene glycol diacetate
Show More Show Less
EINECSNumber 203-881-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.