Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol Diglycidyl Ether (mixture), TCI America™
SDP

Catalog No. E034225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E034225G 25 g
E0342500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E034225G Supplier TCI America Supplier No. E034225G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula C8H14O4
Synonym 1,2-Bis(2,3-epoxypropoxy)ethane, 1,2-Diglycidyloxyethane

Specifications

Color Yellow
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Ethylene Glycol Diglycidyl Ether (mixture)
EINECSNumber (2)-0396&(7)-0343&(2)-0400
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.