Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene glycol, EMPLURA™, MilliporeSigma™

Catalog No. m1009491000
Click to view available options
Quantity:
1 L
Packaging:
Plastic Bottle

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH 6 to 7.5
Melting Point 14°C to 10°C
Density 1.11 g/3 (20 °C)
Boiling Point 197.6°C (1013 hPa)
Quantity 1 L
Flash Point 111°C
Packaging Plastic Bottle
Vapor Pressure 0.053 hPa (20 °C)
Water <0.3%
Solubility Information 1000g/L
Formula Weight 62.07
Percent Purity >99.995%
Show More Show Less
EINECSNumber 203-473-3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.