Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylene Glycol, Fisher BioReagents™

Catalog No. p-25754 Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

Ethylene Glycol

Due to its property as an antifreeze, Ethylene Glycol may be used for the storage of enzymes at low temperatures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH 5.5 to 7.5
Melting Point -13°C
Color Colorless
Decomposition Information >500°C
Density 1.113g/cm³
Boiling Point 196°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Viscous Liquid
Assay Percent Range ≥99 %
Titratable Acid 0.002mEq/g max.
Linear Formula HOCH2CH2OH
Vapor Pressure 0.12mmHg at 20°C
Viscosity 21 cP at 20°C
Water 0.5% max.
Merck Index 15, 3852
Formula Weight 62.06
Percent Purity ≥99%
Chemical Name or Material Ethylene Glycol
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:1
Instability:0
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.