Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylene glycol, Reagent Grade, ≥99%, Honeywell™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| Melting Point | -13°C (literature) |
| CAS Max % | 100.0000% |
| Density | 1.113 g/mL at 25 °C(lit.) |
| Boiling Point | 197°C |
| Quantity | 4 x 2.5 L |
| Flash Point | 111°C |
| Packaging | Poly Bottle |
| Linear Formula | HOCH2CH2OH |
| Beilstein | 505945 |
| Formula Weight | 62.07 |
| Show More |
Safety and Handling
| ShelfLife | 1080 days from date of manufacture |
| EINECSNumber | 203-473-3 |
| Recommended Storage | Room Temp |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction