Ethylene glycol, Spectrophotometric grade, >99%, Alfa Aesar™

Manufacturer: Alfa Aesar 44529K2

Quantity 1L

 View more versions of this product

Catalog No. AA44529K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 107-21-1
    Physical Form Liquid
    Assay Percent Range >99%
    Molecular Formula C2H6O2
    Formula Weight 62.07
    MDL Number MFCD00002885
    Chemical Name or Material Ethylene glycol
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 62.07
    Quantity 1L
    EINECS Number 203-473-3
    Physical Form Liquid
    Density 1.113g/mL
    Assay Percent Range >99%
    Molecular Formula C2H6O2
    MDL Number MFCD00002885
    CAS 107-21-1
    Chemical Name or Material Ethylene glycol
    Grade Spectrophotometric grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit