Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylene/propylene Copolymer, Low Molecular Weight, Thermo Scientific™
Click to view available options
Quantity:
100 g
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Physical Form | Cylindrical Pellets |
Quantity | 100 g |
Infrared Spectrum | Authentic |
Loss on Drying | 2% max. |
Chemical Name or Material | Ethylene/propylene copolymer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction