Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine dihydrochloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
333-18-6
C2H8N2·2HCl
133.04
MFCD00012524
OHHBFEVZJLBKEH-UHFFFAOYSA-N
ethylenediamine dihydrochloride,chlor-ethamine,ethylenediammonium chloride,ethylenediammonium dichloride,ethane-1,2-diamine dihydrochloride,dimethylenediamine dihydrochloride,1,2-diaminoethane dihydrochloride,1,2-ethanediamine, dihydrochloride,ethylenediamine 2hcl,dimethylenediamine diydrochloride
9521
CHEBI:53626
ethane-1,2-diamine;dihydrochloride
C(CN)N.Cl.Cl
333-18-6
C2H8N2·2HCl
133.04
MFCD00012524
OHHBFEVZJLBKEH-UHFFFAOYSA-N
ethylenediamine dihydrochloride,chlor-ethamine,ethylenediammonium chloride,ethylenediammonium dichloride,ethane-1,2-diamine dihydrochloride,dimethylenediamine dihydrochloride,1,2-diaminoethane dihydrochloride,1,2-ethanediamine, dihydrochloride,ethylenediamine 2hcl,dimethylenediamine diydrochloride
9521
CHEBI:53626
ethane-1,2-diamine;dihydrochloride
C(CN)N.Cl.Cl
pH 3.9 to 5 (1% aq. soln.)
Color White
Appearance (1% aq. soln.) clear, colorless
Quantity 2.5kg
Infrared Spectrum Authentic
Melting Point >300.0°C
Packaging Plastic bottle
Assay Percent Range 98+%
Linear Formula H2NCH2CH2NH2·2HCl
Beilstein 04, 230
Merck Index 15, 3849
Solubility Information (1% in water) Clear colorless
Formula Weight 133.04
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material Ethylenediamine dihydrochloride, 98+%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 206-369-6

RTECSNumber : KV3850000

TSCA : TSCA