Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic acid, ACS, 99.4+%

Catalog No. AA1193136
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Chelating agent | CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol

Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
Color White
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 500 g
Beilstein 1716295
Merck Index 14,3517
Solubility Information Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
Formula Weight 292.24
Percent Purity ≥99.4%
Grade ACS
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
Hazard Category H319-H332-H373
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
EINECSNumber 200-449-4
RTECSNumber AH4025000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.