Learn More
Ethylenediaminetetraacetic acid, ACS, 99.4+%
Description
Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | ∼245°C (decomposition) |
Color | White |
Linear Formula | (HO2CCH2)2NCH2CH2N(CH2CO2H)2 |
Quantity | 2.5 kg |
Beilstein | 1716295 |
Merck Index | 14,3517 |
Solubility Information | Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water. |
Formula Weight | 292.24 |
Percent Purity | ≥99.4% |
Grade | ACS |
Show More |
Safety and Handling
Hazard Category | H319-H332-H373 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c |
EINECSNumber | 200-449-4 |
RTECSNumber | AH4025000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.