Learn More
Ethylenediaminetetraacetic acid disodium salt, 0.200N (0.1M) Standardized solution
Description
Ethylenediaminetetraacetic acid disodium salt is widely used in textile industry. Usually applied to dissolve limescale. It is applied in textile industry, pulp and paper industry and also in chelation therapy. In cosmetics, it acts as a sequestering agent. It acts as a corrosion inhibitor to carbon steel in the industries. It also acts as a food additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Colorless |
Odor | Odorless |
Assay Percent Range | 0.200N (0.1M) |
Quantity | 4 L |
Solubility Information | Miscible with water. |
Formula Weight | 336.22 |
Physical Form | Liquid |
Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt |
Safety and Handling
EINECSNumber | 205-358-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.