Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, pure

Catalog No. AC118440025
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118440025 2.5 kg
AC118445000 500 g
AC118440100 10 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC118440025 Supplier Thermo Scientific Chemicals Supplier No. 118440025
Only null left
Add to Cart

Ethylenediaminetetraacetic acid, tetrasodium salt hydrate, 98%, C10H12N2Na4O8.xH2O, CAS Number-194491-31-1 | CAS: 194491-31-1 | C10H12N2Na4O8 | 380.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 194491-31-1
Molecular Formula C10H12N2Na4O8
Molecular Weight (g/mol) 380.17
MDL Number MFCD00150025
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
Synonym edta tetrasodium salt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, tetrasodium 2-2-bis carboxylatomethyl amino ethyl carboxylatomethyl amino acetate hydrate, tetrasodium ion 4-edta hydrate, ethylenediaminetetraacetic acid, tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, bioultra kt, ethylenediaminetetraacetic acid tetrasodium salt hydrate, practical grade
PubChem CID 16211056
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Melting Point >300.0°C
Color White to Yellow
pH 9.5 to 10.5 (1% soln.)
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 97.5% min. (on dry substance) (Complexometry)
Linear Formula (NaO2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O
Fieser 01,373; 05,296
Merck Index 14,3517
Solubility Information Solubility in water: 800g/L (20°C)
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Formula Weight 380.16
Percent Purity 98%
Grade Pure
Infrared Spectrum Authentic
Packaging Plastic bottle
Decomposition Information >400°C
Chemical Name or Material Ethylenediaminetetraacetic acid, tetrasodium salt hydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.