Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid Tetrasodium Salt, Solution, Spectrum™ Chemical
SDP

Catalog No. 18602575
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Drum
Poly Bottle
Poly Drum

E1048, 13235-36-4, C10H12N2O8Na4

Spectrum™ Chemical Ethylenediaminetetraacetic Acid Tetrasodium Salt, Solution is typically known as EDTA and is an aminopolycarboxylic acid. It is usually used to dissolve limescale. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 64-02-8, 7732-18-5
Molecular Formula C10H12N2Na4O8
Molecular Weight (g/mol) 380.17
InChI Key UEUXEKPTXMALOB-UHFFFAOYSA-J
IUPAC Name tetrasodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

CAS Max % 38%, 56%
Packaging Poly Bottle
Quantity 4 L
UN Number UN3267
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.