Learn More
Ethylenediaminetetraacetic acid tripotassium salt
Description
Ethylenediaminetetraacetic acid tripotassium salt is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors. And it is also used to study chelators, biochemicals and reagents. EDTA tripotassium salt has been used in a study to assess the safety of EDTA and HEDTA ingredients in cosmetic formulations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Odor | Odorless |
Quantity | 25 g |
Solubility Information | Soluble in water,0.1 M at 20°C. |
Formula Weight | 406.46 |
Physical Form | Crystalline |
Chemical Name or Material | Ethylenediaminetetraacetic acid tripotassium salt |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 241-543-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.