Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyleoxamate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
617-36-7
C4H7NO3
117.1
MFCD00008005
RZMZBHSKPLVQCP-UHFFFAOYSA-N
ethyl oxamate,ethyl carbamoylformate,oxamic acid ethyl ester,ethyl amino oxo acetate,oxamethane,acetic acid, aminooxo-, ethyl ester,ethoxalamide,oxamidic acid ethyl ester,ethoxy carbonyl formamide,oxalic acid, monoethyl ester amide
69238
ethyl 2-amino-2-oxoacetate
CCOC(=O)C(=O)N
617-36-7
C4H7NO3
117.1
MFCD00008005
RZMZBHSKPLVQCP-UHFFFAOYSA-N
ethyl oxamate,ethyl carbamoylformate,oxamic acid ethyl ester,ethyl amino oxo acetate,oxamethane,acetic acid, aminooxo-, ethyl ester,ethoxalamide,oxamidic acid ethyl ester,ethoxy carbonyl formamide,oxalic acid, monoethyl ester amide
69238
ethyl 2-amino-2-oxoacetate
CCOC(=O)C(=O)N
Melting Point 112°C to 115°C
Quantity 500g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Assay Percent Range 99%
Linear Formula H2NCOCO2C2H5
Beilstein 02, 544
Solubility Information Solubility in water: soluble
Formula Weight 117.1
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity ≥98.5%
Chemical Name or Material Ethyl oxamate
Show More Show Less