Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylgonendione 98.0+%, TCI America™
SDP

Catalog No. E09211G
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 21800-83-9
Molecular Formula C19H26O2
Molecular Weight (g/mol) 286.415
MDL Number MFCD08457890
InChI Key SBLHOJQRZNGHLQ-DIKXUDHVSA-N
Synonym 18-Methylestr-4-en-3,17-dione
PubChem CID 133688345
IUPAC Name (8S,9R,10S,13R,14R)-13-ethyl-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-dione
SMILES CCC12CCC3C(C1CCC2=O)CCC4=CC(=O)CCC34

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 1 g
Formula Weight 286.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylgonendione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.