Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
925-90-6, 109-99-9
C2H5BrMg
133.271
MFCD00000043
FRIJBUGBVQZNTB-UHFFFAOYSA-M
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
101914
magnesium;ethane;bromide
C[CH2-].[Mg+2].[Br-]
925-90-6, 109-99-9
C2H5BrMg
133.271
MFCD00000043
FRIJBUGBVQZNTB-UHFFFAOYSA-M
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
101914
magnesium;ethane;bromide
C[CH2-].[Mg+2].[Br-]
Additional Information magnesium salts may crystallize below 20°C
Linear Formula C2H5MgBr
Concentration 0.85 to 1.15M
Fieser 02,205; 08,510; 11,574
Density 1.0100g/mL
Formula Weight 133.27
Physical Form Solution
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 1.01
Color Black-Brown to Brown
Quantity 800mL
Flash Point −17°C
Chemical Name or Material Ethylmagnesium bromide, 1M Solution in THF
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Harmful if swallowed. May cause respiratory irritation. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. Suspected of causing cancer. Reacts violently with water. May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger