Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylmagnesium bromide, 3M in diethyl ether, AcroSeal™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Density | 1.0300g/mL |
| Flash Point | −40°C |
| Packaging | AcroSeal™ Glass Bottle |
| Linear Formula | C2H5MgBr |
| Quantity | 800 mL |
| Fieser | 02,205 |
| Specific Gravity | 1.03 |
| Formula Weight | 133.27 |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Extremely flammable liquid and vapour. In contact with water releases flammable gases which may ignite spontaneously. Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or di |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove conta |
| EINECSNumber | 213-127-3 |
| TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction