Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmalonic Acid 97%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
601-75-2
C5H8O4
132.115
MFCD00002668
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
ethylmalonic acid,2-ethylmalonic acid,propanedioic acid, ethyl,1,1-propanedicarboxylic acid,alpha-carboxybutyric acid,malonic acid, ethyl,ethylmalonate,ethyl-malonic acid,unii-432nf49dfg,ethylpropanedioic acid
11756
CHEBI:741548
2-ethylpropanedioic acid
CCC(C(=O)O)C(=O)O
601-75-2
C5H8O4
132.115
MFCD00002668
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
ethylmalonic acid,2-ethylmalonic acid,propanedioic acid, ethyl,1,1-propanedicarboxylic acid,alpha-carboxybutyric acid,malonic acid, ethyl,ethylmalonate,ethyl-malonic acid,unii-432nf49dfg,ethylpropanedioic acid
11756
CHEBI:741548
2-ethylpropanedioic acid
CCC(C(=O)O)C(=O)O
Quantity 10g
Color Pink to White
Melting Point 109°C to 114°C
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula HO2CCH(C2H5)CO2H
Beilstein 02, IV, 1952
Formula Weight 132.12
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material Ethylmalonic acid, 97%
Show More Show Less

EINECSNumber : 210-007-2