Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmercury chloride, Thermo Scientific™

Catalog No. aa3711477
Click to view available options
:
0.1g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 107-27-7
Density 3.482g/mL
Molecular Formula C2H5ClHg
Linear Formula CH3CH2HgCl
Quantity 0.1g
MDL Number MFCD00013593
UN Number UN2025
Merck Index 14,3826
Synonym ethylmercury chloride, ceresan, granosan, mercury, chloroethyl, ethylmercuric chloride, cryptodine, ganozan, granozan, emcon d, ethyl mercuric chloride
Solubility Information Insoluble in water; Slightly soluble in ether; Soluble in hot alcohol; Sublimes readily
InChI Key QWUGXIXRFGEYBD-UHFFFAOYSA-M
SMILES CC[Hg]Cl
IUPAC Name chloro(ethyl)mercury
Molecular Weight (g/mol) 265.104
PubChem CID 7862
Formula Weight 265.13
Chemical Name or Material Ethylmercury chloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
RTECSNumber OV9800000
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.