Alfa Aesar™ Ethylphosphonic acid, 98+%

Manufacturer: Alfa Aesar™ B2255914

Quantity 25g

 View more versions of this product

Catalog No. AAB2255914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6779-09-5
    Molecular Formula C2H7O3P
    Formula Weight 110.06
    MDL Number MFCD00002141
    UN Number UN3261
    Beilstein 1743032
    Synonym Ethanephosphonic acid
    Chemical Name or Material Ethylphosphonic acid

    Specifications

    Formula Weight 110.06
    Molecular Formula C2H7O3P
    MDL Number MFCD00002141
    UN Number UN3261
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 25g
    CAS 6779-09-5
    Beilstein 1743032
    Synonym Ethanephosphonic acid
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Melting Point 58° to 62°C
    Chemical Name or Material Ethylphosphonic acid
    Assay ≥98%

    Description

    Applications
    Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

    Solubility
    Soluble in water.

    Notes
    Hygroscopic. Incompatible with strong oxidizing agents and bases.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit