Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylphosphonic acid, 98+%, Thermo Scientific Chemicals

Catalog No. p-7026526
Click to view available options
Quantity:
5 g
25 g

Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylphosphonic acid is used as an internal standard in the determination of fosfomycin in human plasma. It is also used in the synthesis of nucleotide analogs.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6779-09-5
Molecular Formula C2H5O3P
Molecular Weight (g/mol) 108.03
MDL Number MFCD00002141
InChI Key GATNOFPXSDHULC-UHFFFAOYSA-L

Specifications

Melting Point 58°C to 62°C
Quantity 5 g
UN Number UN3261
Beilstein 1743032
Sensitivity Hygroscopic
Synonym phosphonic acid, ethyl, ethylphosphonate, ethanephosphonic acid, phosphate analogue, 6, 4-ethylphosphonic acid, 2-propan-4-ylbenzoic acid, ethylphosphonic acid
Solubility Information Soluble in water.
SMILES CCP(=O)(O)O
IUPAC Name ethylphosphonic acid
PubChem CID 204482
Formula Weight 110.06
Percent Purity ≥98%
Chemical Name or Material Ethylphosphonic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 229-839-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.