Learn More
Ethylurea, 98%
Description
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Ethyl urea, is a versatile building block, that can be used in organic synthesis of various pharmaceutical and biologically active compounds, as nitrogen and carbonyl source.
Solubility
Soluble in methanol (25 mg/ml), and water (100 mg/ml).
Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 88°C to 94°C |
Quantity | 100 g |
Beilstein | 1740512 |
Solubility Information | Soluble in methanol (25mg/ml),and water (100mg/ml). |
Formula Weight | 88.11 |
Percent Purity | 98% |
Chemical Name or Material | Ethylurea |
Safety and Handling
Hazard Category | H302+H312+H332-H335-H341 |
Precautionary Statement | P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c |
EINECSNumber | 210-898-8 |
RTECSNumber | YT2335000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.