Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Eticlopride hydrochloride
GREENER_CHOICESDP

Catalog No. 184750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-475-0 50 mg
18-471-0R 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-475-0 Supplier Tocris Bioscience™ Supplier No. 1847/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

D2 and D3 antagonist (D3 > D2)

Eticlopride hydrochloride is a selective dopamine D2/D3 receptor antagonist (Ki values are 0.50 and 0.16 nM respectively). Antipsychotic.

Specifications

Chemical Name or Material 3-Chloro-5-ethyl-N-[[(2S)-1-ethyl-2-pyrrolidinyl)methyl]-6-hydroxy-2-methoxy-benzamide hydrochloride
CAS 97612-24-3
Assay Percent Range >99%
Quantity 50 mg
Solubility Soluble to 100mM in water
Molecular Formula C17H25ClN2O3.HCl
Formula Weight Observed MW: 377.31

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.