Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETK Monoclonal Antibody (1C6)
GREENER_CHOICE

Catalog No. PIMA515634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515634 Supplier Invitrogen™ Supplier No. MA515634
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15634 targets BMX in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15634 immunogen is purified recombinant fragment of human BMX expressed in E. Coli.. MA5-15634 detects BMX which has a predicted molecular weight of approximately 78kDa.

Bone marrow tyrosine kinase (BMX) is a non-receptor tyrosine kinase that belongs to the SRC-related TEC subfamily of tyrosine kinases. It is an important regulator of various cellular processes including apoptosis, cell survival, cellular differentiation, cell migration, and transformation. One family of nonreceptor TKs includes the genes TEC (MIM 600583), TXK (MIM 600058), ITK (MIM 186973), and BTK (MIM 300300). All of these proteins are homologs of the Drosophila Src28 TK and contain an SH3 and SH2 domain upstream of the TK domain.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene BMX
Gene Accession No. P51813
Gene Alias BMX; BMX non-receptor tyrosine kinase; bone marrow tyrosine kinase gene in chromosome X protein; Bone marrow tyrosine kinase gene in chromosome X protein homolog; BTK-like on X chromosome; cytoplasmic tyrosine-protein kinase BMX; epa3; Epithelial and endothelial tyrosine kinase; ETK; Etk/Bmx; Etk/Bmx cytosolic tyrosine kinase; NTK38; NTK38 tyrosine kinase; PSCTK2; PSCTK3; RGD1565643; Tyro8; TYRO8 protein tyrosine kinase 8
Gene Symbols BMX
Host Species Mouse
Immunogen Purified recombinant fragment of human BMX expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 660
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.