Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETNK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583086
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583086 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583086 Supplier Invitrogen™ Supplier No. PA583086
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EFIQGEALDP KHVCNPAIFR LIARQLAKIH AIHAHNGWIP KSNLWLKMGK YFSLIPTGFA DEDINKRFLS DIPSSQILQE EMTWMKEILS NLGSPVVLCH NDLLCKNIIY NE Highest antigen sequence indentity to the following orthologs: Rat - 96%, Mouse - 94%.

Ethanolamine kinase 1 (EKI1), functions in the first committed step of the phosphatidylethanolamine synthesis pathway. This cytosolic enzyme is specific for ethanolamine and exhibits negligible kinase activity on choline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETNK1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ETNK1
Gene Accession No. Q9HBU6
Gene Alias 1110061E11Rik; 4930555L11Rik; AI195356; AI841245; C80956; D6Ertd3e; EKI; EKI 1; EKI1; Ethanolamine kinase; Ethanolamine kinase 1; Etnk1; I79_004753; Nbla10396; putative protein product of Nbla10396
Gene Symbols ETNK1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ETNK1. Recombinant protein control fragment (Product #RP-90291).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.