Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETNK2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563410
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563410 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563410 Supplier Invitrogen™ Supplier No. PA563410
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFHLRRHTPC PQCSWGMEEK AAASASCREP PGPPRAAAVA YFGISVDPDD ILPGALRLIQ EL Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 76%.

The protein encoded by this gene is a member of choline/ethanolamine kinase family which catalyzes the first step of phosphatidylethanolamine (PtdEtn) biosynthesis via the cytidine diphosphate (CDP) ethanolamine pathway. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Jul 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETNK2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ETNK2
Gene Accession No. Q9NVF9
Gene Alias 4933417N20Rik; AI197444; EKI 2; EKI2; ethanolamine kinase 2; Ethanolamine kinase-like protein; ETNK2; HMFT1716; putative ethanolamine kinase; Tuc1
Gene Symbols ETNK2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ETNK2. Recombinant protein control fragment (Product #RP-99803).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55224
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.