Learn More
R&D Systems™ Etoposide
SDP
Catalog No. 468409606 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
46-840-9606 100 μL
1 options
This item is not returnable. View return policy
Catalog No. 46-840-9606 Supplier R&D Systems™ Supplier No. 468409606
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
R&D Systems™ Etoposide complexes with topoisomerase II and DNA to enhance doublestrand and single-strand cleavage of DNA and reversibly inhibit religation.
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
  • Do not use past expiration date.
  • Blocks the cell cycle in S-phase and G2-phase; induces apoptosis in normal and tumor cell lines; inhibits synthesis of the oncoprotein Mdm2 and induces apoptosis in tumor lines that overexpress Mdm2.
  • Etoposide is provided in DMSO. The final reagent concentration is 10mM.
Specifications
Quantity 100 μL
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Product Line R&D Systems