Learn More
Trajan™ ETP Active Film™ Electron Multipliers
SDP
Drop-in, plug-compatible replacement electron multipliers
Supplier:  Trajan™ 14823H
 View more versions of this product
Catalog No. SG14823H
Description
Description
  • Detect presence of ion signals emerging from the mass analyzer of mass spectrometer; also reduce multiplier noise and increase signals
  • Available for all major instruments including Agilent Technologies™ Perkin-Elmer™, Shimadzu™, Thermo Scientific, and Varian™; total compatibility with GC/MS, LC/MS, ICP/MS, TOF/MS, and Magnetic Sector techniques
  • Features Active Film™—proprietary dynode material with a chemically durable metal oxide secondary emission system; very air stable; exhibits stable performance even after repeated exposure to atmosphere
  • Large dynode surface area for extended operating life
  • High operational reliability and mechanical robustness; designed for sensitivity and linear system response
  • Optimized ion and electron optics for maximum performance
  • Easy installation
  • Meet EPA, DFTPP ion abundance criteria
  • Two-year shelf life
Specifications
Specifications
Electron Multiplier
Sensar/Larson-Davis TOF 2000 TOF Analyzer Type MS
Large Dynode
EPA, DFTPP
TOF/MS - Detect Presence of Ion Signals Emerging From The Mass Analyzer of Mass Spectrometer, Also Reduce Multiplier Noise and Increase Signals
662°F
350°C
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
ShelfLife : 2 Years
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.