Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Ets-1 Ab-1, Mouse Monoclonal Antibody

Catalog No. MS1762R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS1762R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1762R1 Supplier Epredia™ Supplier No. MS1762R1

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Ets-1 Ab-1, Mouse Monoclonal Antibody.

Ets-1 is a transcription factor involved in the regulation of genes required for extracellular matrix remodeling during metastasis of tumors. It is known to interact with the enhancer of urokinase like plasminogen activator, and promoters of stromelysin-1 and collagenase-1. It is absent from normal gastric mucosa but is expressed in 60% of gastric carcinoma and oral squamous cell carcinoma.

Host Species: Mouse

Clone: 1G11

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: aa 226-454

Immunogen: Recombinant oncoprotein corresponding to aa 226-454 of human Ets-1

Molecular Weight: 51 kDa

Positive Control: Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Ets-1 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1G11
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant oncoprotein corresponding to aa 226-454 of human Ets-1
Quantity 1 mL
Regulatory Status RUO
Research Discipline Transcription Regulation
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.