Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ets-1 Rabbit anti-Human, Rat, Clone: 5P4K1, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315807100UL
This item is not returnable.
View return policy
Description
Ets-1 Monoclonal antibody specifically detects Ets-1 in Human, Rat samples. It is validated for Western BlotSpecifications
| Ets-1 | |
| Monoclonal | |
| Unconjugated | |
| Avian erythroblastosis virus E26 (v-ets) oncogene homolog-1, ets protein, ETS-1, EWSR2, FLJ10768, p54, protein C-ets-1, v-ets avian erythroblastosis virus E2 oncogene homolog 1, v-ets avian erythroblastosis virus E26 oncogene homolog 1, v-ets erythroblastosis virus E26 oncogene homolog 1 (avian) | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Ets-1 (P14921). | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Chromatin Research, Innate Immunity, Transcription Factors and Regulators | |
| 2113 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 5P4K1 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction