Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione Peroxidase 3/GPX3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications
$215.00 - $518.50
Specifications
| Antigen | Glutathione Peroxidase 3/GPX3 |
|---|---|
| Dilution | Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 24167362), Immunoblotting reported in scientific literature (PMID 29571702) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP106398SS
![]() |
Novus Biologicals
NBP106398SS |
0.025 mL |
Each for $215.00
|
|
|||||
NBP106398
![]() |
Novus Biologicals
NBP106398 |
0.1 mL |
Each for $518.50
|
|
|||||
Description
Glutathione Peroxidase 3/GPX3 Polyclonal specifically detects Glutathione Peroxidase 3/GPX3 in Human, Rat, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.Specifications
| Glutathione Peroxidase 3/GPX3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Lipid and Metabolism, Signal Transduction | |
| Whole antisera with 0.1% Sodium Azide | |
| EC 1.11.1, EC 1.11.1.9, Extracellular glutathione peroxidase, glutathione peroxidase 3, glutathione peroxidase 3 (plasma), GPx-3, GPx-P, GPXP, GSHPx-3, GSHPx-P, Plasma glutathione peroxidase | |
| GPX3 | |
| IgG | |
| Unpurified | |
| Human, monkey and rat. Immunogen sequence has 89% identity to mouse, 88% identity to bovine and 84% identity to porcine. |
| Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 24167362), Immunoblotting reported in scientific literature (PMID 29571702) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Rat, Primate | |
| P22352 | |
| 2878 | |
| Full length human Glutathione Peroxidase 3 expressed in E. coli. [Swiss-Prot# P22352] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 25 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title