Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS2 Antibody (OTI2A3), DyLight 755, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270694IR

Catalog No. NBP270694IR


Only null left
Add to Cart

Description

Description

ETS2 Monoclonal specifically detects ETS2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

ETS2
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
ETS2
Recombinant protein fragment corresponding to amino acids 90-349 of human ETS2 (NP_005230) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI2A3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian)
Mouse
Affinity Purified
Stem Cell Markers
2114.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.