Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETS2 Mouse anti-Human, Clone: PCRP-ETS2-1D9, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313877100UG
This item is not returnable.
View return policy
Description
ETS2 Monoclonal antibody specifically detects ETS2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Protein ArraySpecifications
ETS2 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, ELISA, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Protein Array | |
P15036 | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation, Peptide Array | |
PCRP-ETS2-1D9 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian) | |
Recombinant full-length human ETS2 protein (Uniprot: P15036) | |
100 μg | |
Stem Cell Markers | |
2114 | |
Store at 4°C. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction