Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ETS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01674426
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-426 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-426 Supplier Invitrogen™ Supplier No. PA19543
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

PA1-9543 detects ETS2 in human samples. PA1-9543 has been successfully used in Western blot procedures. By Western blot, PA1-9543 detects a 55 kDa protein representing ETS2. PA1-9543 immunogen is a purified recombinant ETS2.3 corresponding to the chromosome 21 gene (Human).

ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ETS2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ETS2
Gene Accession No. P15036
Gene Alias AU022856; avian erythroblastosis virus E26 (v-ets) oncogene homolog 2; E26 avian leukemia oncogene 2, 3' domain; ETS proto-oncogene 2, transcription factor; Ets2; Ets-2; ETS2IT1; oncogene ETS-2; protein C-ets-2; v-ets avian erythroblastosis virus E2 oncogene homolog 2; v-ets avian erythroblastosis virus E26 oncogene homolog 2; v-ets erythroblastosis virus E26 oncogene homolog 2
Gene Symbols ETS2
Host Species Chicken
Immunogen Purified recombinant ETS2.3 corresponding to the chromosome 21 gene (Human).
Purification Method IgY fraction
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2114
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.